Learn More
Gibco™ FluoroBrite™ DMEM

Description
Gibco™ FluoroBrite™ DMEM features a background fluorescence that is comparable to PBS and 90% lower than that emitted by standard phenol red–free DMEM. Formulated to include the required nutrients for routine cell culture when supplemented with 10% fetal bovine serum and 4 mM L-glutamine or GlutaMAX™ Supplement, FluoroBrite™ DMEM is designed to enhance the signal-to-noise ratio of fluorophores, enabling researchers to visualize even the weakest fluorescent events in an environment that promotes optimum cell health. Additional features include:
- Enhancement of fluorescence signal during live-cell imaging
- DMEM-based to help preserve cell health
Live-cell fluorescence microscopy is an essential technique for the visualization of fundamentally important and physiologically relevant biological events. A key challenge with this technique is the ability to image weak fluorophores without causing cell damage, photobleaching, or undesirable changes to cell health. FluoroBrite™ DMEM helps address these issues.
Specifications
Specifications
| Classification | Animal Origin-free |
| Concentration | 1 X |
| Form | Liquid |
| Product Type | DMEM (Dulbecco's Modified Eagle Medium) |
| Serum Level | Standard Serum Supplementation |
| Sterility | Sterile-filtered |
| With Additives | High Glucose |
| Without Additives | No Glutamine, No Phenol Red, No HEPES, No Sodium Pyruvate |
| Manufacturing Quality | cGMP-compliant under the ISO 13485 standard |
| Quantity | 10 x 500 mL |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.