Learn More
Gibco™ FluoroBrite™ DMEM
Gibco™ FluoroBrite™ DMEM features a background fluorescence that is comparable to PBS and 90% lower than that emitted by standard phenol red–free DMEM.
Brand: Gibco™ A1896701
33.65 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.60000kg
Description
Gibco™ FluoroBrite™ DMEM features a background fluorescence that is comparable to PBS and 90% lower than that emitted by standard phenol red–free DMEM. Formulated to include the required nutrients for routine cell culture when supplemented with 10% fetal bovine serum and 4 mM L-glutamine or GlutaMAX™ Supplement, FluoroBrite™ DMEM is designed to enhance the signal-to-noise ratio of fluorophores, enabling researchers to visualize even the weakest fluorescent events in an environment that promotes optimum cell health. Additional features include:
• Enhancement of fluorescence signal during live-cell imaging
• DMEM-based to help preserve cell health
Live-cell fluorescence microscopy is an essential technique for the visualization of fundamentally important and physiologically relevant biological events. A key challenge with this technique is the ability to image weak fluorophores without causing cell damage, photobleaching, or undesirable changes to cell health. FluoroBrite™ DMEM helps address these issues.
Specifications
Store in refrigerator (2–8°C). Protect from light. | |
1 X | |
DMEM (Dulbecco's Modified Eagle Medium) | |
Sterile-filtered | |
No Glutamine, No HEPES, No Phenol Red, No Sodium Pyruvate | |
cGMP-compliant under the ISO 13485 standard |
Animal Origin-free | |
Liquid | |
Standard Serum Supplementation | |
High Glucose | |
Sustainable packaging | |
500 mL |
For Research Use Only. Not for use in diagnostic procedures.