Learn More
Fluorocyclohexane, 97%, Thermo Scientific Chemicals
£100.00 - £300.00
Chemical Identifiers
CAS | 372-46-3 |
---|---|
Molecular Formula | C6H11F |
Molecular Weight (g/mol) | 102.15 |
InChI Key | GOBGVVAHHOUMDK-UHFFFAOYSA-N |
Synonym | cyclohexane, fluoro, cyclohexyl fluoride, cyclohexylfluoride, fluoro-cyclohexane, acmc-1cpcv |
PubChem CID | 78988 |
IUPAC Name | fluorocyclohexane |
SMILES | C1CCC(CC1)F |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
372-46-3 | |
102.15 | |
cyclohexane, fluoro, cyclohexyl fluoride, cyclohexylfluoride, fluoro-cyclohexane, acmc-1cpcv | |
fluorocyclohexane |
C6H11F | |
GOBGVVAHHOUMDK-UHFFFAOYSA-N | |
78988 | |
C1CCC(CC1)F |
Specifications
372-46-3 | |
0.9280g/mL | |
5°C | |
96% min. (GC) | |
C6H11F | |
cyclohexane, fluoro, cyclohexyl fluoride, cyclohexylfluoride, fluoro-cyclohexane, acmc-1cpcv | |
C1CCC(CC1)F | |
102.15 | |
102.15 | |
Fluorocyclohexane |
13.0°C | |
103.0°C to 105.0°C | |
Authentic | |
C6H11F | |
0.928 | |
GOBGVVAHHOUMDK-UHFFFAOYSA-N | |
fluorocyclohexane | |
78988 | |
97% |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 206-754-9
- Fluorocyclohexane
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only