Learn More
Cytiva Sample loops
Fitted FEP tubing sample loops. Cytiva Sample loops supplied with ferrules and unions.
Brand:  Cytiva 18-1161-24
Product Code. 11340042
  • £158.00 / Each
Edge
Explore more special offers
Description
Description
  • Tubing i.d. 1.6 mm (0.063 inch) o.d. 1/8 in. fitted with nut 5/16 in. male and ferrules
  • 2 x Union UNF 5/16 in. female-1/16 in. male supplied with the sample loop
  • Fluoroethenepropene (FEP)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 mL