missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fluoromount-G™ Mounting Medium
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
Description
- Fluoromount-G™ does not fluoresce and may reduce the amount of fluorochrome quenching during fluorescence microscopy
- Clear liquid medium
- Supplied as a 1x stock solution containing sodium azide
Specifications
Specifications
| Quantity | 25 mL |
| Product Type | Mounting Medium |
| Product Line | Fluoromount-G |
| Reagent Type | Mounting Solution |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction