missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fluoroskan Ascent™ Microplate Fluorometer

Perform a variety of research applications with the compact, robust Thermo Scientific™ Fluoroskan Ascent™ Microplate Fluorometer.

Specifications

Depth (Metric) 42cm
Detector Type Photomultiplier Tube
Incubator Temperature Ambient +3° to 45°C
Plate Type 1- to 384-well plates
Shaking Orbital
View More Specs

Description

Description

Perform a variety of research applications with the compact and robust Thermo Scientific™ Fluoroskan Ascent™ Microplate Fluorometer. Featuring excellent optical performance, this microplate fluorometer is ideal for life science research applications such as fluorometric protein and enzyme studies, molecular interactions, nucleic acid quantification, reporter gene, fluorometric kinase, immuno and cell based assays.

Onboard dispensers for exact follow-up of kinetic reactions

  • Comes equipped with up to three reagent dispensers for fast kinetic assays, such as Ca2+ flux
  • Supports simultaneous dispensing and reading, enabling monitoring of fast kinetic measurements from the very start of the reaction
  • Allows conservation of expensive reagents with very low dead volume and the backflush capability

High sensitivity for both top and bottom reading

  • Fiberless direct illumination optics for both top and bottom reading
  • Ensures a high sensitivity, a wide dynamic range and accurate and precise results for 1- to 384-well plates

Specially designed for automation

  • Reads up to 384-well plates
  • Easily integrates with automated systems
  • Enables the most rapid throughput

Includes:

PC Software and filter pairs: Excitation: 355nm/Emission: 460nm, Excitation: 485nm/ Emission: 538nm

Recommended for:

Ca2+ flux assays; Cell proliferation; Cytotoxicity; Multi-drug resistance; Cell adhesion; DNA quantitation; Reporter gene assays; Hybridization assays; Ion Channel; Quantitation of PCR products; FRET assays; Molecular beacon assays; Immunoassays; Enzyme activity; Neonatalogy; Bacterial quantitation; Phagocytosis; Oligonucleotide assays; ADMEtox

Warranty

1 year

Specifications

Specifications

42cm
Ambient +3° to 45°C
Orbital
>6 decades
360 to 800nm
320 to 700nm
34cm
Serial RS-232C port
2fmol fluorescein/well in a black 96-well plate
46 lb.
16.5 in.
Photomultiplier Tube
1- to 384-well plates
Filters
Up to eight filters in the emission filter wheel. 460 and 538nm filters included as standard.
Up to eight filters in the excitation filter wheel. 355 and 485nm filters included as standard.
13.4 in.
PC Software and filter pairs: Excitation: 355nm/Emission: 460nm, Excitation: 485nm/ Emission: 538nm
Quartz-halogen lamp
One Year
21kg
42cm
Videos
SDS
Documents

Documents

Product Certifications