Learn More
Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 410715000
Code : 29
Additional Details : CAS Number : 16961-83-4 Weight : 0.50000kg Transport : UN number : 1778 Chem class : 8 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Fluosilicic acid | |
16961-83-4,7732-18-5 | |
Plastic bottle | |
500 g | |
Liquid | |
MFCD00036289 | |
OHORFAFFMDIQRR-UHFFFAOYSA-P | |
hexafluorosilicon(2-);hydron | |
21863527 | |
19 to 20.6% (total fluorine) |
Colorless | |
1.2700g/mL | |
1.27 | |
F6H2Si | |
H2SiF6 | |
hexafluorosilicic acid, fluorosilicic acid, dihydrogen hexafluorosilicate, silicate 2-, hexafluoro-, dihydrogen, hexafluorosilicon 2-; hydron, dihydrogen hexafluorosilanediuide, silicofluoridd, hexafluorosilic acid, hydrofluorosilic acid, kieselfluorwasserstoffsaure | |
[H+].[H+].F[Si-2](F)(F)(F)(F)F | |
144.091 | |
144.09 |
Safety and Handling
- Fluosilicic acid, 25 wt.% aqueous solution
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only