missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ FluoSpheres™ Size Kit #1, Carboxylate-modified Microspheres, red fluorescent (580/605), 2% solids, six sizes

FluoSpheres™ Size Kit #1, Carboxylate-modified Microspheres, red fluorescent (580/605), 2% solids, six sizes

Brand:  Molecular Probes™ F8887

Code : 50

Additional Details : Weight : 0.00450kg

Product Code. 10583663

  • £866.00 / 1 set
Estimated Shipment: 05-08-2024
to see stock.

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene. Our Molecular Probes™ FluoSpheres™ beads are manufactured using high-quality, ultraclean polystyrene and are loaded with a variety of our proprietary dyes to create intensely fluorescent beads that typically show little or no photobleaching, even when excited with the intense illumination required for fluorescence microscopy. This FluoSpheres™ Size Kit supplies 1 mL of each the 0.02, 0.1, 0.2, 0.5, 1.0, and 2.0μm sizes.

FluoSpheres™ Microsphere Specifications
  • Label (Ex/Em): Red fluorescent (580/605)
  • Nominal bead diameter: 0.02, 0.1, 0.2, 0.5, 1.0, 2.0μm
  • Coupling surface: Carboxylate
  • Solids: 2%

    Characteristics of the Various FluoSpheres™ Coupling Surfaces
  • Carboxylate-modified FluoSpheres™ beads have a high density of pendent carboxylic acids on their surface, making them suitable for covalent coupling of proteins and other amine-containing biomolecules using water-soluble carbodiimide reagents such as EDAC.
  • Sulfate FluoSpheres™ beads are relatively hydrophobic particles that will passively and nearly irreversibly adsorb almost any protein, including albumin, IgG, avidin, and streptavidin.
  • Aldehyde—sulfate FluoSpheres™ beads have surface aldehyde groups added, designed to react with proteins and other amines under very mild conditions.
  • Amine-modified FluoSpheres™ beads can be coupled to a wide variety of amine-reactive molecules, including the succinimidyl esters and isothiocyanates of haptens and drugs or the carboxylic acids of proteins, using a water-soluble carbodiimide.

    Key Applications of Microspheres
  • Instrument calibration (flow cytometry, microscopy, HTS, HCS)
  • Flow testing (microfluidics, blood flow, water flow, and air flow)
  • Cell biology tracers (cell differentiation and cell tracing)
  • Immunoassays (agglutination tests, ELISA, particle capture, and contrast reagents)

    Choices for FluoSpheres™ Fluorescent Microspheres
    Among our complete offering of fluorescent microspheres products, you’ll find beads with these variations:
  • Ten fluorescent colors
  • Ten nominal bead diameters: 0.02μm, 0.04μm, 0.1μm, 0.2μm, 0.5μm, 1.0μm, 2.0μm, 4.0μm, 10.0μm, and 15.0μm
  • Four surface modifications for protein coupling: carboxylate, sulfate, aldehyde-sulfate, amine
  • Microspheres that are additionally precoupled with streptavidin, NeutrAvidin, biotin, europium, and platinum

    Choices for Unstained Microspheres
    We also offer hundreds of choices for UltraClean™ surfactant-free microspheres for research and commercial applications.

    We’ll Make a Custom Microsphere Product for You
    We will prepare custom orders upon request. For example, FluoSpheres™ beads can be prepared with intensities that are lower than those of our regular selection, a desirable feature in some multicolor applications. Our custom conjugation service is efficient and confidential, and we guarantee the quality of our work. We are ISO 9001:2000 certified.

    For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
  • Specifications

    Specifications

    0.02μm: 2.3 x 1015 Particles/mL, 0.1μm: 3.6 x 1013 Particles/mL, 0.2μm: 4.6 x 1012 Particles/mL, 0.5μm: 3.1 x 1011 Particles/mL, 1μm: 3.8 x 1010 Particles/mL, 2μm: 5.3 x 109 Particles/mL
    Distilled water, 2mM azide
    Red Fluorescent
    Polystyrene
    6 x 1 mL
    0.02, 0.1, 0.2, 0.5, 1, 2 μm
    2°C to 8°C
    Carboxylate Modified Microsphere Kit #1, 6 x 1mL, Six Sizes: 0.02 μm, 0.1 μm, 0.2 μm, 0.5 μm, 1 μm, 2 μm, Red, 2% Solid, Microspheres (Also Called Latex Beads or Latex Particles) are Spherical Particles in the Colloidal Size Range That a
    Instrument Calibration (Flow Cytometry, Microscopy, HTS, HCS), Flow Testing (Microfluidics, Blood Flow, Water Flow, and Air Flow), Cell Biology Tracers (Cell Differentiation and Cell Tracing), Immunoassays (Agglutination Tests, ELISA, Particle Capture)
    1.055g/cm3
    Carboxylate
    0.02, 0.1, 0.2, 0.5, 1, 2 μm
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use Only. Not for use in diagnostic procedures.