Learn More
FLYWCH2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559531
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SKDSTKVAGA KRKGVHCVMS LGVPGPATLA KALLQTHPEA QRAIEAAPQE PEQKRSRQDP GTDRTEDSGL AAGPPEAAGE NFA Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 70%.
FLYWCH2 is a protein coding gene.Specifications
FLYWCH2 | |
Polyclonal | |
Unconjugated | |
FLYWCH2 | |
FLYWCH family member 2; FLYWCH2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114984 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96CP2 | |
FLYWCH2 | |
Recombinant protein corresponding to Human FLYWCH2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only