missing translation for 'onlineSavingsMsg'
Learn More

FMOC-L-Isoleucine, 99%, Thermo Scientific™

Product Code. 10125110
Click to view available options
Quantity:
2.5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Melting Point 146°C to 149°C
CAS 71989-23-6
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C21H23NO4
Packaging Glass Bottle
Linear Formula FMOC-NHCH(COOH)CH(CH3)C2H5
MDL Number MFCD00037125
Specific Rotation -11° to -13° (20°C, 589nm) (c=1, DMF)
Synonym fmoc-ile-oh, fmoc-l-isoleucine, n-fmoc-l-isoleucine, fmoc-l-ile-oh, n-9h-fluoren-9-ylmethoxy carbonyl-l-isoleucine, n-9-fluorenylmethoxycarbonyl-l-isoleucine, fmoc-ile, fmoc-l-ile, 2s,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylpentanoic acid, 9-fluorenylmethoxycarbonyl-l-isoleucine
InChI Key QXVFEIPAZSXRGM-DJJJIMSYSA-N
SMILES CC[C@H](C)[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
IUPAC Name (2S,3S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylpentanoic acid
Specific Rotation Condition - 12.00 (20.00°C c=1,DMF)
Molecular Weight (g/mol) 353.42
Quantity 2.5 g
PubChem CID 2724629
Formula Weight 353.42
Percent Purity ≥98.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material FMOC-L-Isoleucine, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'