Learn More
Applied Biosystems™ Fmoc-L-Phenylalanine, 10/Pk.
These are Amino Acid Cartridges for the ABI™ 431-433 Peptide Synthesizer.
Brand: Applied Biosystems™ 410645
Additional Details : Weight : 0.04540kg
Description
- For Research Use Only
- Not for use in diagnostics procedures
Specifications
Each package contains 10 cartridges. | |
433 A Peptide Synthesizer | |
Peptide Synthesis Reagents |
Cartridge | |
Protein Sequencing & Peptide Synthesis | |
10 Each |