missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Fmoc-PAL-PEG-PS™
Shop All Applied Biosystems Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
Description
For Research Use Only. Not for use in diagnostics procedures.
Specifications
Specifications
| For Use With (Equipment) | 433 A Peptide Synthesizer |
| Product Line | PEG-PS |
| Content And Storage | Store in refrigerator (2°C to 8°C). |
| Product Type | Resin |
| Quantity | 5 g |
| Format | Resin |
| Shipping Condition | Room Temperature |
| Matrix | Polyethylene glycol - polystyrene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction