missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FN3K Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528603
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2. Predicted reactivity: Mouse (91%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FN3K catalyzes phosphorylation of fructosamines formed by glycation, the nonenzymatic reaction of glucose with primary amines followed by Amadori rearrangement. Phosphorylation of fructosamines may initiate metabolism of the modified amine and result in deglycation of glycated proteins.Specifications
FN3K | |
Polyclonal | |
Unconjugated | |
Fn3k | |
2310074G21Rik; FN3K; Fnsk; fructosamine 3 kinase; fructosamine 3-kinase; fructosamine-3-kinase; Protein-psicosamine 3-kinase FN3K; Protein-ribulosamine 3-kinase FN3K | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64122 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.94 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9H479 | |
Fn3k | |
Recombinant fragment corresponding to a region within amino acids 1 and 309 of Human FN3K | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |