missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ FN3KRP 293T Cell Transient Overexpression Lysate (Denatured)

Human overexpression lysate

Brand:  Abnova™ H00079672-T01.100uL


Product Code. 16164333

  • £182.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

293T
FN3KRP 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 34.1kDa
Transient overexpression cell lysate was tested with anti-FN3KRP antibody (Mfr. No. H00079672-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
NM_024619.2
pCMV-FN3KRP full-length
100 μL
Human
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 100UL FN3KRP 293T Cell Transient Overexpression Ly

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contains : Mercaptoethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers