Learn More
Tocris Bioscience™ FNIR-Tag, NHS
Near-infrared fluorescent dye for labeling of amines; supplied as NHS ester
Brand:  Tocris Bioscience™ 7373/2
Additional Details : Weight : 0.00970kg
Product Code. 18769296
  • £322.89 / 2mg
Estimated Shipment: 08-07-2024
to see stock.
Description
Description
FNIR-Tag-NHS is a near-infrared fluorescent dye; supplied as an NHS ester for coupling to primary amine groups. FNIR-Tag-NHS allows a higher degree of labeling (DOL) with less aggregation than IRDye™ 800CW. When conjugated to antibodies, FNIR-Tag-NHS exhibits reduced liver uptake in vivo compared with IRDye™ 800CW conjugates. In addition, FNIR-Tag-NHS conjugates are sigificantly brighter than IRDye™ 800CW conjugates at similar labeling densities in vivo and in vitro. Antibody conjugation efficiency is greatest at pH 7.4. Excitation/emission maxima = 766/788 nm; Extinction coefficient = 200,000 M-1cm-1; QY = 0.099.
IRDye™ is a registered trademark of LI-COR Biosciences.
Specifications
Specifications
2-((E)-2-((E)-2-(3-((4-((2,5-dioxopyrrolidin-1-yl)oxy)-4-oxobutyl)dimethylammonio)propoxy)-3-(2-((E)-1-(2-(2-(2-methoxyethoxy)ethoxy)ethyl)-3,3-dimethyl-5-sulfonatoindolin-2-ylidene)ethylidene)cyclohex-1-en-1-yl)vinyl)-1-(2-(2-(2-methoxyethoxy)ethoxy)ethy
2 mg
C57H80N4O17S2
2365033-54-9
Fluorescent Dyes
0.9