missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Folic acid dihydrate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cellsFolic acid dehydrate is used to form non-covalent complexes with polyethylenimine. It is used for the delivery of genes, which finds application in gene therapy. It is a B-complex vitamin, which is necessary for the body to produce red blood cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cellsFolic acid dehydrate is used to form non-covalent complexes with polyethylenimine. It is used for the delivery of genes, which finds application in gene therapy. It is a B-complex vitamin, which is necessary for the body to produce red blood cells.
Solubility
Miscible with water.
Notes
Light sensitive. Incompatible with strong oxidizing agents.
Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cellsFolic acid dehydrate is used to form non-covalent complexes with polyethylenimine. It is used for the delivery of genes, which finds application in gene therapy. It is a B-complex vitamin, which is necessary for the body to produce red blood cells.
Solubility
Miscible with water.
Notes
Light sensitive. Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∼250°C (decomposition) |
Quantity | 25 g |
Beilstein | 100781 |
Merck Index | 14,4221 |
Solubility Information | Miscible with water. |
Optical Rotation | +18° (c=0.5 in 0.1N NaOH) |
Formula Weight | 477.44 (441.40 Anhydrous) |
Percent Purity | 97% |
Chemical Name or Material | Folic acid dihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'