Learn More
SCHOTT AG Lighting and Imaging™ Footswitch for VisiLED Series
This Footswitch for VisiLED Series enables the user to remotely switch between stored memory positions.
Brand: SCHOTT AG Lighting and Imaging™ 400020
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Foot pedal quickly scrolls from 1 through 4 to change the stored illumination conditions.
- Allows the user to quickly and easily change to various illumination types by allowing total hand control under the microscope.
- For use with the VisiLED Controller MC1500.
Specifications
Footswitch | |
Stereo Microscopy Illumination | |
VisiLED Controller MC1500 |