Learn More
3M™ Forestry Headgear Combination
3M™ Forestry Headgear Combination provides a versatile and comfortable solution for wearers who require both face and hearing protection.
Brand: 3M™ 7000103790
565.33 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- A practical, convenient solution for face and hearing protection where no head protection is needed
- Front cover provides ventilation for increased comfort and also designed as a cable holder for combination with communication products
- Mesh Face Shield: + 5°C (Stainless Steel) is approved for High Speed Particles 45 m/s (F) according to EN1731
- To increase eye protection, G500 can incorporate the 3M™ V6 integrated safety glasses range, that are available in clear, gray and yellow lenses
- Headgear can be used both with and without earmuffs
- Headband is slotted for the attachment of earmuffs
Specifications
Forestry | |
Headgear Combination |