Learn More
Thermo Scientific™ Forma™ Freezer Replacement Shelves
Use Thermo Scientific™ Forma™ Freezer Replacement Shelves with Forma 8600 and 900 Series Upright Freezers.
Brand:  Thermo Scientific™ 224134
 View more versions of this product
Product Code. 11786469
  • £171.21 / Each
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Use Thermo Scientific™ Forma™ Freezer Replacement Shelves with Forma 8600 and 900 Series Upright Freezers.
  • Stainless steel
  • Kits include mounting hardware
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Shelf kit
13 cu. ft.
Upright
13.0 cu. ft. freezers
Shelf Kit
368.1 L
Silver