missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Forma™ Freezer Replacement Shelves
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
13.0 cu. ft. Freezers
17.3 cu. ft. Freezers
23.0 cu. ft. Freezers
28 cu. ft. Freezers
Type:
Shelf Kit
Unit Size:
Each
Description
Use Thermo Scientific™ Forma™ Freezer Replacement Shelves with Forma 8600 and 900 Series Upright Freezers.
- Stainless steel
- Kits include mounting hardware
Specifications
Specifications
| Type | Shelf Kit |
| Capacity (English) | 28 cu. ft. |
| Capacity (Metric) | 792.8 L |
| Style | Upright |
| Color | Silver |
| For Use With (Equipment) | 28 cu. ft. Freezers |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction