Learn More
Formal Buffered Saline, Reagecon
Fixative mixture of formaldehyde in isotonic saline for routine histopathology.
Brand: Reagecon FBS5
Code : NEW
Additional Details : CAS Number : 50-00-0 Weight : 5.00000kg Transport : UN number : 3267 Chem class : 8 Pack group : III
Description
- Fixative for histopathology.
Specifications
Liquid | |
HDPE Jerrycan | |
5 L |
Safety and Handling
- FORMAL BUFFERED SALINE
Signal Word
- Danger
Hazard Category
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H317-May cause an allergic skin reaction.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
- MIXTURE LIST-Contains : Formaldehyde, Sodiumdihydrogenphosphate