missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Formal-Fixx™

Product Code. p-4532383 Shop All Epredia Products
Click to view available options
Quantity:
3.8 L concentrate (19 L total)
Product Type:
Fixative
Unit Size:
3.8 L
This item is not returnable. View return policy

Product Code. 10415625

Brand: Epredia™ 6764255

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Acquire amazing results with Epredia™ Formal-Fixx™, which is excellent for immunochemistry.

  • Enhances H&Es and special stains
  • Minimizes the need to enzyme digest
  • Reduces protein cross-linking
  • Produces well-defined nuclear detail
TRUSTED_SUSTAINABILITY

Specifications

Product Type Fixative
Quantity 3.8 L concentrate (19 L total)
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
  • H370-Causes damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : formaldehyde, zinc sulphate, methyl alcohol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.