missing translation for 'onlineSavingsMsg'
Learn More
Formaldehyde (37% by Weight/Molecular Biology), Fisher BioReagents Product Code.: 10459113

Formaldehyde (37% by Weight/Molecular Biology), Fisher BioReagents

Product Code. 10459113
25 mL, Glass Bottle
Click to view available options
Quantity:
25 mL
500 mL
Packaging:
Glass Bottle
HDPE Bottle
This item is not returnable. View return policy

Product Code. 10459113

Brand: Fisher BioReagents BP53125

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Molecular Biology-grade Formaldehyde is used for denaturing RNA.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-00-0, 67-56-1, 7732-18-5
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Melting Point −15°C
Color Colorless
Density 1.083g/cm┬│
pH 3 to 4.2
Boiling Point 97°C
Ignition Residue 0.005% max.
Assay Percent Range 36.5 to 38.0 %
Packaging Glass Bottle
Linear Formula HCHO
Quantity 25 mL
Merck Index 15, 4263
Formula Weight 30.02
Vapor Pressure 2mbar at 20┬░C
Percent Purity 36.5 to 38.0%
Grade Molecular Biology
Physical Form Liquid
Chemical Name or Material Formaldehyde, 37% by Weight
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formaldehyde, solution 37%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity after single exposure Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H370-Causes damage to organs.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Formaldehyde, Methylalcohol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Formaldehyde (37% by Weight/Molecular Biology), Fisher BioReagents >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.