missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde (37% by Weight/Molecular Biology), Fisher BioReagents
Click to view available options
Quantity:
25 mL
500 mL
Packaging:
Glass Bottle
HDPE Bottle
Description
Molecular Biology-grade Formaldehyde is used for denaturing RNA.

Specifications
Specifications
Melting Point | −15°C |
Color | Colorless |
Density | 1.083g/cm┬│ |
pH | 3 to 4.2 |
Boiling Point | 97°C |
Ignition Residue | 0.005% max. |
Assay Percent Range | 36.5 to 38.0 % |
Packaging | HDPE Bottle |
Linear Formula | HCHO |
Quantity | 500 mL |
Show More |
Safety and Handling




Product Identifier
- Formaldehyde, solution 37%
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Specific target organ toxicity after single exposure Category 1
- H226-Flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H370-Causes damage to organs.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Formaldehyde, Methylalcohol
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Formaldehyde (37% by Weight/Molecular Biology), Fisher BioReagents >
Spot an opportunity for improvement?Share a Content Correction