missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol

Formaldehyde, formalin, CAS # 50-00-0, is a colorless volatile used as a histologic fixative and disinfectant.

Brand:  Thermo Scientific Chemicals 410731000

16.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.



 View more versions of this product

Product Code. 10299980

  • £20.06 / 100g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Formaldehyde (formalin) is an organic compound used for crosslinking proteins in biological samples such as tissue sections, culture cells, and whole specimens
  • Formaldehyde is an intermediate in the combustion of methane and can exist in many forms
  • It is a volatile, colorless liquid; can be stored long-term in appropriate containers
  • Comes stabilized in solution with methanol

Applications

  • May be used in pain research for induction of pain and inflammation
  • Monomeric formaldehyde is formulated especially for laboratory applications
  • Commonly used in RNA isolation as a denaturant
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468720 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number410730050,410731000,410730010
Lot NumberA0466472 Certificate TypeCertificate of Analysis Date01/10/2024 Catalog Number410730050,410730010,410731000
Lot NumberA0465755 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog Number410730050,410730010,410731000
Lot NumberA0465756 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog Number410730050,410730010,410731000
Lot NumberB0547598A Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number410730050,410730010,410731000
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol > 100g; Plastic bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only