missing translation for 'onlineSavingsMsg'
Learn More
Formaldehyde, Analytical Grade, Penta Chemicals Product Code.: 30020545
compliance-icons compliance-icons compliance-icons

Formaldehyde, Analytical Grade, Penta Chemicals

Product Code. 30020545
3 L
Click to view available options
Quantity:
3 L
This item is not returnable. View return policy

Product Code. 30020545

Brand: PENTA CHEMICALS 1415020003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Top quality chemicals for laboratories, schools, hospitals, research institutes, and large manufacturing plants.

About us

The company PENTA was established in 1990 under the trade name Ing. Petr Švec - PENTA s.r.o.. From the very beginning, we have been a solely Czech company which is managed by the Švec family, including company founder Petr Švec.

We specialize in the manufacture and distribution of pure, laboratory, pharmaceutical and specialty chemicals, which we market under the trade name Penta. We manufacture and supply a wide range of organic and inorganic chemicals, in large volumes, with a quality ranging from technical, to pure, to A.G. and ACS.

High quality and flexibility

Thanks to the high quality of our products, and our ability to fulfil specific customer requirements, we rank among the leading suppliers of qualified chemicals in the domestic market, and more recently we have also expanded into the global market.

We manufacture the chemicals in our own modern complex, which fulfils the strictest quality and safety criteria. Our large warehouse facilities, including two automated warehouses, enable us to accomodate client requirements. We package the products in accordance with all legislative requirements, and can deliver them to the customer using our own ADR transport on request.

Environmental responsibility

Our priority is to supply the maximum quality of chemicals, reliability, and environmental awareness. Our efforts are evidenced by many awards and certificates, including ISO 9001, ISO 14001 and the Responsible Care certificate for responsible business in the chemical industry.

Chemical Identifiers

Molecular Weight (g/mol) 30.03

Specifications

Quantity 3 L
Percent Purity 36 to 38%
Grade Analytical
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formaldehyde 36 - 38%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H371-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : Formaldehyde, methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Formaldehyde, Analytical Grade, Penta Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.