missing translation for 'onlineSavingsMsg'
Learn More
Formaldehyde Solution Saline, Phosphate Buffered pH7.3, Pure, Fisher Chemical Product Code.: 10040170

Formaldehyde Solution Saline, Phosphate Buffered pH7.3, Pure, Fisher Chemical

Product Code. 10040170
25 L
Click to view available options
Quantity:
25 L
This item is not returnable. View return policy

Product Code. 10040170

Brand: Fisher Chemical F/1515/25

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Melting Point −5°C/23°F
Color Clear
pH 7.3
Boiling Point 100°C
Packaging Plastic container
MDL Number 146963
Quantity 25 L
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Formaldehyde solution saline , phosphate buffered pH 7,3
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H371-May cause damage to organs.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Formaldehyde Solution Saline, Phosphate Buffered pH7.3, Pure, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.