missing translation for 'onlineSavingsMsg'
Learn More
Formamide (≥99.5%), Fisher BioReagents Product Code.: 10523525

Formamide (≥99.5%), Fisher BioReagents

Product Code. 10523525
100 mL, Amber Glass
Click to view available options
Quantity:
100 mL
Packaging:
Amber Glass
This item is not returnable. View return policy

Product Code. 10523525

Brand: Fisher BioReagents BP228100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For nucleic acid hybridization and sequencing

Super Pure Formamide has been pretreated with a mixed-bed resin so that it is ready for use in nucleic acid hybridization and sequencing.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 2°C
Absorbance 0.05 max. at 280nm
CAS 75-12-7
Density 1.133g/cm³
Boiling Point 210°C
Quantity 100 mL
Identification Pass Test
Packaging Amber Glass
Physical Form Liquid
Assay Percent Range ≥99.5 %
Molecular Formula CH3NO
Linear Formula H2NCH:O
MDL Number MFCD00007941
Vapor Pressure 0.08mbar at 20°C
Viscosity 3.75 mPa/s at 20°C
Merck Index 15, 4265
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
SMILES NC=O
IUPAC Name formamide
Molecular Weight (g/mol) 45.04
PubChem CID 713
ChEBI CHEBI:48431
Formula Weight 45.04
Percent Purity ≥99.5%
Grade Biotech
Chemical Name or Material Formamide
Show More Show Less
compliance-icons
Product Identifier
  • Formamide
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Formamide (≥99.5%), Fisher BioReagents >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.