missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formamide, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
500 mL
2.5 L
25 L
Packaging:
Plastic Container
Plastic-coated Glass Bottle
Description
Formamide, Certified AR for Analysis
Specifications
Specifications
pH | 4 to 5 |
Melting Point | 2°C |
Boiling Point | 210°C |
Quantity | 25 L |
Packaging | Plastic Container |
Physical Form | Liquid |
Vapor Pressure | 0.08mbar at 20°C |
Viscosity | 3.75 mPaS at 20°C |
Formula Weight | 45.04g/mol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'