missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formamide (Molecular Biology), Fisher BioReagents
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
Description
Molecular Biology-grade Formamide is commonly used in nucleic acid hybridization and sequencing. It requires pretreatment with a mixed-bed resin.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
pH | 4 to 5 |
Melting Point | 2°C |
Absorbance | 0.1 max. at 280nm |
Density | 1.133g/cm³ |
Boiling Point | 210°C |
Quantity | 100 mL |
Identification | Pass Test |
Packaging | Glass Bottle |
Physical Form | Liquid |
Assay Percent Range | ≥99 % |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Formamide
- Danger
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- H351-Suspected of causing cancer.
- H360FD-May damage fertility. May damage the unborn child.
- H373-May cause damage to organs.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'