missing translation for 'onlineSavingsMsg'
Learn More

Formamide, puriss. p.a., ACS Reagent, ≥99.5% (GC/T), Honeywell Riedel-de Haën™

Puriss. p.a., ACS Reagent,≥99.5% (GC/T)

£50.31 - £196.03

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15645650
View Documents
Honeywell-Riedel-de Haen
47670-4X25ML
4 x 25 mL Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15635650
View Documents
Honeywell-Riedel-de Haen
47670-250ML
250 mL Glass Bottle
£50.31
250mL
Please to purchase this item. Need a web account? Register with us today!
15615650
View Documents Promotion Details
Honeywell-Riedel-de Haen
47670-1L
1 L Glass Bottle
£88.22
1 L
Please to purchase this item. Need a web account? Register with us today!
15625650
View Documents
Honeywell-Riedel-de Haen
47670-2.5L
2.5 L Glass Bottle
£196.03
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Chemical Identifiers

75-12-7
45.04
ZHNUHDYFZUAESO-UHFFFAOYSA-N
713
formamide
CH3NO
MFCD00007941
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
CHEBI:48431
NC=O

Specifications

4 to 5 (at 20°C)
75-12-7
1.132g/mL at 20°C, 1.134g/mL at 25°C
175°C (347°F)
Weak
CH3NO
MFCD00007941
NONH for all modes of transport
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
NC=O
45.04
CHEBI:48431
≥99.5% (GC/T)
1°C to 3°C
Colorless
210°C
932°F
Liquid
HCONH2
30mmHg (129°C), 0.08mmHg (20°C)
505995
ZHNUHDYFZUAESO-UHFFFAOYSA-N
formamide
713
45.04g/mol
Formamide
Videos
Safety and Handling

Safety and Handling

P201-P260-P280-P308 + P313

missing translation for 'rtecsNumber' : 200-842-0

Product Identifier
  • Formamide

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications