missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formamide, Ultra Pure, 99.5% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
250 mL
Description
Formamide is used in the synthesis of medicine, animal glue and other organic compound. It is also used as a solvent which ionizes water-insoluble compounds. It is used as a softening agent for paper and fiber. It is widely used in the fields of the spice, dye, plastic as well as textile industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Formamide is used in the synthesis of medicine, animal glue and other organic compound. It is also used as a solvent which ionizes water-insoluble compounds. It is used as a softening agent for paper and fiber. It is widely used in the fields of the spice, dye, plastic as well as textile industries.
Solubility
Soluble in water (20 mg/ml ), methanol, ethanol and acetone. Slightly soluble in ether.
Notes
Hygroscopic. Store at room temperature. Store away from water or moisture.
Formamide is used in the synthesis of medicine, animal glue and other organic compound. It is also used as a solvent which ionizes water-insoluble compounds. It is used as a softening agent for paper and fiber. It is widely used in the fields of the spice, dye, plastic as well as textile industries.
Solubility
Soluble in water (20 mg/ml ), methanol, ethanol and acetone. Slightly soluble in ether.
Notes
Hygroscopic. Store at room temperature. Store away from water or moisture.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 2°C |
Color | Colorless |
Density | 1.134 g/mL |
Boiling Point | 210°C |
Quantity | 100 mL |
Flash Point | 150°C (302°F) |
Odor | Ammonia-like |
Physical Form | Liquid |
Refractive Index | 1.4475 |
Beilstein | 505995 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'