missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formamidine acetate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Formamidine acetate is used as an intermediate in the synthesis of active pharmaceutical ingredients. It is also used as a condensing agent for the preparation of pyrimidine and imidazole heterocycles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Formamidine acetate is used as an intermediate in the synthesis of active pharmaceutical ingredients. It is also used as a condensing agent for the preparation of pyrimidine and imidazole heterocycles.
Solubility
Soluble in water and hot alcohol.
Notes
Hygroscopic. Moisture sensitive. Incompatible with oxidizing agents.
Formamidine acetate is used as an intermediate in the synthesis of active pharmaceutical ingredients. It is also used as a condensing agent for the preparation of pyrimidine and imidazole heterocycles.
Solubility
Soluble in water and hot alcohol.
Notes
Hygroscopic. Moisture sensitive. Incompatible with oxidizing agents.
Specifications
Melting Point | ∼158°C (decomposition) |
Density | 1.124 |
pH | 8 |
Quantity | 100 g |
Beilstein | 3563488 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water and hot alcohol. |
Formula Weight | 104.11 |
Percent Purity | 99% |
Chemical Name or Material | Formamidine acetate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'