missing translation for 'onlineSavingsMsg'
Learn More

Formamidine acetate, 99%, Thermo Scientific Chemicals

Product Code. 11459083
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Formamidine acetate is used as an intermediate in the synthesis of active pharmaceutical ingredients. It is also used as a condensing agent for the preparation of pyrimidine and imidazole heterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Formamidine acetate is used as an intermediate in the synthesis of active pharmaceutical ingredients. It is also used as a condensing agent for the preparation of pyrimidine and imidazole heterocycles.

Solubility
Soluble in water and hot alcohol.

Notes
Hygroscopic. Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3473-63-0
Molecular Formula C3H8N2O2
Molecular Weight (g/mol) 104.109
MDL Number MFCD00012866
InChI Key XPOLVIIHTDKJRY-UHFFFAOYSA-N
Synonym formamidine acetate, methanimidamide, monoacetate, formamidinium acetate, acetic acid; methanimidamide, formamidine acetate salt, formamidine, monoacetate, formimidamide acetate, imidoformamide acetate, acetic acid; formamidine, methanimidamide monoacetate
PubChem CID 160693
IUPAC Name acetic acid;methanimidamide
SMILES CC(=O)O.C(=N)N

Specifications

Melting Point ∼158°C (decomposition)
Density 1.124
pH 8
Quantity 500 g
Beilstein 3563488
Sensitivity Hygroscopic
Solubility Information Soluble in water and hot alcohol.
Formula Weight 104.11
Percent Purity 99%
Chemical Name or Material Formamidine acetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.