Learn More
Formamidinesulfinic acid, 99%, Thermo Scientific Chemicals
£18.70 - £74.20
Chemical Identifiers
CAS | 1758-73-2 |
---|---|
Molecular Formula | CH4N2O2S |
Molecular Weight (g/mol) | 108.11 |
MDL Number | MFCD00002397 |
InChI Key | FYOWZTWVYZOZSI-UHFFFAOYSA-N |
Synonym | thiourea dioxide, formamidinesulfinic acid, amino imino methanesulfinic acid, formamidine sulfinic acid, manofast, thiourea s,s-dioxide, aimsa, methanesulfinic acid, aminoimino, carbamimidoylsulfinic acid, methenesulfinic acid, aminoimino |
PubChem CID | 61274 |
IUPAC Name | amino(imino)methanesulfinic acid |
SMILES | C(=N)(N)S(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10466920
|
Thermo Scientific Acros
119720100 |
10 g | Glass Bottle |
£18.70
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10666112
|
Thermo Scientific Acros
119721000 |
100 g | Plastic bottle |
£21.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10172910
|
Thermo Scientific Acros
119725000 |
500 g | Plastic bottle |
£74.20
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1758-73-2 | |
108.11 | |
FYOWZTWVYZOZSI-UHFFFAOYSA-N | |
61274 | |
C(=N)(N)S(=O)O |
CH4N2O2S | |
MFCD00002397 | |
thiourea dioxide, formamidinesulfinic acid, amino imino methanesulfinic acid, formamidine sulfinic acid, manofast, thiourea s,s-dioxide, aimsa, methanesulfinic acid, aminoimino, carbamimidoylsulfinic acid, methenesulfinic acid, aminoimino | |
amino(imino)methanesulfinic acid |
Specifications
1758-73-2 | |
White | |
99% | |
H2NC(=NH)SO2H | |
03, I, 36 | |
thiourea dioxide, formamidinesulfinic acid, amino imino methanesulfinic acid, formamidine sulfinic acid, manofast, thiourea s,s-dioxide, aimsa, methanesulfinic acid, aminoimino, carbamimidoylsulfinic acid, methenesulfinic acid, aminoimino | |
FYOWZTWVYZOZSI-UHFFFAOYSA-N | |
amino(imino)methanesulfinic acid | |
61274 | |
99% | |
Formamidinesulfinic acid, 99% |
126.0°C | |
Authentic | |
CH4N2O2S | |
MFCD00002397 | |
04,506; 05,668; 06,586 | |
Solubility in water: 30g/L (20°C). Other solubilities: insoluble in organic solvents | |
C(=N)(N)S(=O)O | |
108.11 | |
108.11 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Self-heating in large quantities; may catch fire.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.
May cause damage t
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep cool.
Protect from sunlight.
Avoid breathing dust/fume/ga
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 217-157-8
missing translation for 'tsca' : TSCA
- Formamidinesulfinic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Self-heating substance Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H373-May cause damage to organs.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H252-Self-heating in large quantities; may catch fire.
Precautionary Statement
- P235+P410-Keep cool. Protect from sunlight.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only