Learn More
Formic acid, 85%, Thermo Scientific Chemicals
£25.93 - £51.66
Chemical Identifiers
CAS | 64-18-6 |
---|---|
Molecular Formula | CH2O2 |
Molecular Weight (g/mol) | 46.025 |
MDL Number | MFCD00003297 |
InChI Key | BDAGIHXWWSANSR-UHFFFAOYSA-N |
Synonym | methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglattShow More |
PubChem CID | 284 |
ChEBI | CHEBI:30751 |
IUPAC Name | formic acid |
SMILES | C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11338406
|
Thermo Scientific Alfa Aesar
L17434.0B |
1000 g |
£25.93
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11406850
|
Thermo Scientific Alfa Aesar
L17434.0E |
2500 g |
£51.66
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Formic acid is principally used as a preservative antibacterial agent in livestock feed, in dying textiles, tanning leather and electroplating. In the poultry industry, it is sometimes added with feed to kill E. coli bacteria. It acts as a reducing agent for the catalytic reduction of nitrate in water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFormic acid is principally used as a preservative antibacterial agent in livestock feed, in dying textiles, tanning leather and electroplating. In the poultry industry, it is sometimes added with feed to kill E. coli bacteria. It acts as a reducing agent for the catalytic reduction of nitrate in water.
Solubility
Miscible with water, ether, acetone, ethyl acetate, glycerol, methanol and ethanol. Partially miscible with benzene, toluene and xylene.
Notes
Hygroscopic. Store closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
Chemical Identifiers
64-18-6 | |
46.025 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
284 | |
formic acid |
CH2O2 | |
MFCD00003297 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglattShow More | |
CHEBI:30751 | |
C(=O)O |
Specifications
64-18-6 | |
100°C to 101°C | |
HCOOH | |
UN1779 | |
14,4241 | |
Miscible with water,ether,acetone,ethyl acetate,glycerol,methanol and ethanol. Partially miscible with benzene,toluene and xylene. | |
C(=O)O | |
46.025 | |
CHEBI:30751 | |
85% | |
50°C (156°F) | |
Hygroscopic |
7°C to 9°C | |
CH2O2 | |
MFCD00003297 | |
1209246 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
formic acid | |
284 | |
46.03 | |
1.22 | |
1.371 | |
Formic acid |
Safety and Handling
GHS H Statement
H331-H314-H318-H226-H302
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P312-P330-P331-P363-P372-P374-P380-P501c
H226-H302-H314-H331
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: FORMIC ACID
missing translation for 'einecsNumber' : 200-579-1
missing translation for 'rtecsNumber' : LQ4900000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Formic acid, 85%
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
- P331-Do NOT induce vomiting.
- P363-Wash contaminated clothing before reuse.
- P372-Explosion risk in case of fire.
- P374-Fight fire with normal precautions from a reasonable distance.
- P380-Evacuate area.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P404-Store in a closed container.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : formic acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.