missing translation for 'onlineSavingsMsg'
Learn More
Formic acid, 98+%, pure, Thermo Scientific Chemicals Product Code.: 10071620

Formic acid, 98+%, pure, Thermo Scientific Chemicals

Product Code. 10071620
1 L, Plastic bottle
Click to view available options
Quantity:
25 mL
250 mL
1 L
2.5 L
10 L
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10071620

Brand: Thermo Scientific Acros 147930010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Assay Percent Range 98+%
Linear Formula HCO2H
Beilstein 02, 8
Fieser 01,404; 02,202; 03,147; 04,239; 05,316; 07,160; 08,232; 09,226; 11,243; 13,107; 16,177
Merck Index 15, 4269
Formula Weight 46.02
Percent Purity ≥98.0%
Grade Pure
Density 1.2200g/mL
Quantity 1 L
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Liquid
Refractive Index 1.3701 to 1.3721 (20°C, 589nm)
Vapor Pressure 44mbar at 20°C
Chemical Name or Material Formic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0460391 Certificate TypeCertificate of Analysis Date21/02/2024 Catalog Number147932500,147930025,147930100,147930010,147930250
Lot NumberB0541429A Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number147932500,147930025,147930100,147930010,147930250
Lot NumberB0539961A Certificate TypeCertificate of Analysis Date20/10/2023 Catalog Number147932500,147930025,147930100,147930010,147930250
Lot NumberB0539389A Certificate TypeCertificate of Analysis Date20/10/2023 Catalog Number147932500,147930025,147930100,147930010,147930250
Lot NumberA0438424 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number147932500,147930025,147930100,147930010,147930250
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Formic acid, 98+%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.