missing translation for 'onlineSavingsMsg'
Learn More

Formic acid ≥99%, BAKER ANALYZED™ LC-MS Reagent, J.T. Baker™

Product Code. 15508664 Shop All J.T.Baker Products
Click to view available options
Quantity:
2 mL
10 mL
Unit Size:
2mL
Each
This item is not returnable. View return policy

Product Code. 15508664

Brand: JT Baker 9820.0002AM

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tested for UHPLC-MS

  • LC-Gradient with Diode Array Detection (AU), 0.1% (v/v) aqueous solution:
    • at 220 nm max. 0.002
    • at 254 nm max. 0.002
  • LC-MS Gradient Suitability Test (TIC, 100 to 2000 m/z), 0.1% (v/v) aqueous solution:
    • Positive ESI-MS Sensitive Impurities (as Reserpine) max. 50 ppb
  • Ultraviolet Absorbance, 0.1% (v/v) aqueous solution (1.00-path vs water):
    • at 240 nm max. 0.1
    • at 260 nm max. 0.01

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.03
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym Methanoic acid 100%
IUPAC Name formic acid
SMILES OC=O

Specifications

Assay Percent Range min. 99%
UN Number 1779
Quantity 2 mL
Flash Point 69°C
Residue after Evaporation max. 20 ppm
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.