missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, ACS Reagent, 88 to 91%, Honeywell™ Fluka™

Product Code. 15645390
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent, 88-91%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

pH Acidic
Melting Point 4°C
Boiling Point ∼100.4°C
Color Colorless
Appearance Liquid
Linear Formula HCOOH
UN Number UN1779
Beilstein 1209246
Solubility Information Water soluble
Formula Weight 46.03g/mol
Percent Purity 88 to 91%
Density 1.22g/mL at 25°C
Quantity 500 mL
Explosion Limit 12% to 38%
Flash Point 48°C (118.4°F)
Ignition Point 1004°F
Odor Stinging
Packaging Poly Bottle
Refractive Index n20/D 1.370
Vapor Density 1.6 (vs air)
Vapor Pressure 44.8mmHg (20°C)
Chemical Name or Material Formic acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'