missing translation for 'onlineSavingsMsg'
Learn More

Formic acid-d2, for NMR, +99% atom D, 95% solution in D2O, Thermo Scientific Chemicals

Product Code. 10742194
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920-42-3
Molecular Formula CH2O2
Molecular Weight (g/mol) 48.037
MDL Number MFCD00037363
InChI Key BDAGIHXWWSANSR-PFUFQJKNSA-N
Synonym formic acid-d2, 2h formic 2 acid, dcood, 2h2 formic acid, formic-d acid-d 6ci,7ci,8ci,9ci, formic-d acid-d, 95 wt% in deuterium oxide, formic acid-d2, 95 wt. % in d2o, 98 atom % d
PubChem CID 123092
IUPAC Name deuterio deuterioformate
SMILES C(=O)O

Specifications

Physical Form Liquid
Color Colorless
Density 1.2700g/mL
Deuterium (D) 99.0% min.
Flash Point 50°C
Packaging Glass Bottle
Specific Gravity 1.27
Quantity 1 g
Melting Point 5.9°C
Boiling Point 101.0°C
Linear Formula DCOOD
Solubility Information Solubility in water: miscible.
Formula Weight 48.04
Percent Purity ≥94.0%
Grade NMR
Chemical Name or Material Formic acid-d2, 99+ atom % D (95% in D2O)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formic acid-d2
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'