missing translation for 'onlineSavingsMsg'
Learn More

Formic acid, Eluent additive for LC-MS, Honeywell Fluka™

Eluent additive for LC-MS

£92.05 - £902.75

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15658430
View Documents
Honeywell-Fluka
56302-10X1ML
10 x 1 mL Ampoule
£92.05
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
15665840
View Documents
Honeywell-Fluka
56302-50ML
50 mL Glass Bottle
£766.00
50mL
Please to purchase this item. Need a web account? Register with us today!
15675840
View Documents
Honeywell-Fluka
56302-50ML-GL
50 mL Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15655840
View Documents
Honeywell-Fluka
56302-1L-GL
1 L Glass Bottle
£902.75
1 L
Please to purchase this item. Need a web account? Register with us today!
15645840
View Documents
Honeywell-Fluka
56302-1L
1 L Plastic Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

64-18-6
46.025
BDAGIHXWWSANSR-UHFFFAOYSA-N
284
formic acid
CH2O2
MFCD00003297
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
CHEBI:30751
C(=O)O

Specifications

64-18-6
100.4°C
≤100.0000%
H2O
UN1779
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
C(=O)O
46.025
CHEBI:30751
>97.5%
1.22 g/mL at 25 °C(lit.)
49.5°C
1.6 (vs air)
Formic acid, Eluent additive for LC-MS
8.2°C to 8.4°C (literature)
≥90.0000%
CH2O2
MFCD00003297
1209246
BDAGIHXWWSANSR-UHFFFAOYSA-N
formic acid
284
46.03
LC-MS
0.57
1004°F
44.8 mmHg (20°C)
Videos
Safety and Handling

Safety and Handling

P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P403 + P233

missing translation for 'shelfLife' : 1080 days from date of manufacture

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Formic acid

missing translation for 'einecsNumber' : 200-579-1

missing translation for 'rtecsNumber' : 200-579-1

Recommended Storage : Room Temp

Product Identifier
  • Formic acid

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • EUH071-Corrosive to the respiratory tract.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications