missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic acid, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥98%, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥98%
£40.12 - £750.51
Chemical Identifiers
CAS | 64-18-6 |
---|---|
Molecular Formula | CH2O2 |
Molecular Weight (g/mol) | 46.025 |
MDL Number | MFCD00003297 |
InChI Key | BDAGIHXWWSANSR-UHFFFAOYSA-N |
Synonym | methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt |
PubChem CID | 284 |
ChEBI | CHEBI:30751 |
IUPAC Name | formic acid |
SMILES | C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15633870
|
Honeywell-Fluka
33015-500ML |
500 mL |
£40.12
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15603870
|
Honeywell-Fluka
33015-1L |
1 L |
£65.25
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15613870
|
Honeywell-Fluka
33015-2.5L |
2.5 L |
£127.69
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15653870
|
Honeywell-Fluka
33015-6X1L |
6 x 1 L | N/A | ||||||
15623870
|
Honeywell-Fluka
33015-4X2.5L |
4 x 2.5 L | N/A | ||||||
15663870
|
Honeywell-Fluka
33015-70KG |
70 kg | N/A | ||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
64-18-6 | |
46.025 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
284 | |
formic acid |
CH2O2 | |
MFCD00003297 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
CHEBI:30751 | |
C(=O)O |
Specifications
64-18-6 | |
HCOOH | |
1779 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
C(=O)O | |
46.025 | |
CHEBI:30751 | |
≥98% | |
1.22g/mL at 25°C | |
49°C (120.2°F) | |
1.6 (vs air) | |
Formic Acid, Reag. Ph. Eur. |
CH2O2 | |
MFCD00003297 | |
1209246 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
formic acid | |
284 | |
46.03g/mol | |
ACS/puriss. p.a. | |
0.57 | |
1004°F | |
44.8mmHg (20°C) |
Safety and Handling
P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P403 + P233
H226-H302-H314-H331
missing translation for 'einecsNumber' : 200-579-1
missing translation for 'rtecsNumber' : LQ4900000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Formic acid puriss. ACS reagent
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.