missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic Acid, TraceSELECT™, ≥88.0% (T), Honeywell™ Fluka™
Click to view available options
Quantity:
250 mL
Packaging:
Glass bottle
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
pH | Acidic |
CAS | 64-18-6 |
Melting Point | 4°C |
Boiling Point | ∼100.4°C (1013hPa) |
Color | Colorless |
Molecular Formula | CH2O2 |
Linear Formula | HCOOH |
MDL Number | MFCD00003297 |
UN Number | UN1779 |
Beilstein | 1209246 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'