missing translation for 'onlineSavingsMsg'
Learn More

Forskolin, (from Coleus forskohlii), MP Biomedicals™

Product Code. 11456071
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 66575-29-9
Molecular Formula C22H34O7
Molecular Weight (g/mol) 410.507
MDL Number MFCD00082317
InChI Key OHCQJHSOBUTRHG-KGGHGJDLSA-N
Synonym forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish
PubChem CID 47936
ChEBI CHEBI:42471
IUPAC Name [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate
SMILES CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O

Specifications

Color White
Assay Percent Range 0.9775
Quantity 5 mg
Formula Weight 410.5
Physical Form Powder
Chemical Name or Material Forskolin, from Coleus forskohlii
compliance-icons
Product Identifier
  • Forskoli
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.