Learn More
Poulten Graf™ Fortuna™ Optima™ All Glass Luer-Tip Syringe
Excellent for pharmaceutical, cosmetics and biotechnological industries. Poulten Graf™ Fortuna™ Optima™ All Glass Luer-Tip Syringes come with an interchangeable piston.
Specifications
Autoclavable Autoclavable
Certifications/Compliance CE EN 46001
Material Borosilicate Glass
Description Amber graduation
Product Type All Glass Luer-tip Syringe
View More Specs
Description
Description
  • Complies with CE EN 46001 standards
  • Autoclavable at 134°C
  • Constructed from Durobax™ glass
  • With permanent amber graduation
  • Acid and alkali resistant
Specifications
Specifications
Autoclavable
Borosilicate Glass
All Glass Luer-tip Syringe
Luer Tip
CE EN 46001
Amber graduation
1/Ea.
Yes
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Poulten Graf™ Fortuna™ Optima™ All Glass Luer-Tip Syringe