Learn More
Cole Parmer Fowler Digital Micrometer
Fowler Digital Micrometer features a thimble ratchet and lock.
Brand:  Cole Parmer 97231-61
Product Code. 11795449
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Fowler Digital Micrometer features a thimble ratchet and lock.
  • Provides instant readings to 0.00005 in. on the 6-digit LCD.
  • Zero preset
  • Hold
  • Relative/absolute function
  • Auto power on and off
  • Carbide tipped anvils
  • Includes 1.5 V battery, fitted storage case, and wrench for calibration.
  • Range 0 to 1 in. (0 to 25 mm)
Specifications
Specifications
Ruler