Learn More
Cole Parmer Fowler Micrometer Calibration Set
Fowler Micrometer Calibration Set is ideal for maintenance personnel and technicians, as well as beginner and professional users.
Brand:  Cole Parmer 97236-02
Product Code. 11789789
  • £414.00 / Each
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Fowler Micrometer Calibration Set is ideal for maintenance personnel and technicians, as well as beginner and professional users.
  • A ring gauge is used to measure, verify and/or calibrate gauge diameter sizes.
  • Ring gauge kits are offered in sizes ranging from 0.0625 in. up to 4 in. and are available with standard steel blocks or for abrasion and corrosion resistance choose zirconium-based ceramic blocks.
  • For the most accurate measurements, NIST-traceable calibration services for your gauge blocks are provided.
  • Calibration Gauge Block Kits are available with steel blocks or zirconium-based ceramic blocks that are abrasion- and corrosion-resistant.
  • Kits include a protective wooden case, and NIST traceable Certificate
Specifications
Specifications
Ruler