missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXC1 Mouse anti-Human, Clone: 1F4E11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66568-1-IG-150UL
This item is not returnable.
View return policy
Description
FOXC1 is a protein belonging to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this protein is not known, however, it plays a role in the regulation of FGF19-FGFR4-MAPK pathway to promote both the development and maintece of anterior segment structures within the eye. Mutations in this gene cause various glaucoma, iridogoniodysgenesis anomaly, Peters anomaly that includes central corneal leukoma, absence of the posterior corneal stroma and Descemetmembrane and Axenfeld-Rieger anomaly characterized by posterior corneal embryotoxon, iris adhesion to the Schwalbe line, hypertelorism, hypoplasia of the malar bones, congenital absence of some teeth and mental retardation.Specifications
FOXC1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q12948 | |
FOXC1 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
1F4E11 | |
Unconjugated | |
FOXC1 | |
ARA, FKHL7, forkhead box C1, Forkhead box protein C1, Forkhead related protein FKHL7, FOXC1, FREAC 3, FREAC3, IGDA, IHG1, IRID1, RIEG3 | |
Mouse | |
Protein G | |
RUO | |
2296 | |
-20°C | |
Liquid |