missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxP3 Mouse anti-Human, BB700, Clone: 236A/E7, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566526
This item is not returnable.
View return policy
Description
FoxP3 Monoclonal antibody specifically detects FoxP3 in Human samples. It is validated for Flow Cytometry.Specifications
FoxP3 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
100 Tests | |
Primary | |
Human | |
IgG1 κ |
Flow Cytometry | |
236A/E7 | |
Mouse | |
RUO | |
FoxP3 | |
Refrigerate (2 to 8°C) |