missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva FR-902 FFKM / PEEK / Stainless Steel Flow Restrictor
Used when using low pressure columns and/or when the pH electrode is mounted in the pH flow cell. Cytiva FR-902 FFKM / PEEK / Stainless Steel Flow Restrictor is pre-set to 0.2 MPa.
Brand: Cytiva 18-1121-35
Code : GX
Additional Details : Weight : 0.02160kg
Description
- The wetted parts are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, except 100 % Ethyl acetate, 100 % Hexane and 100 % Tetrahydrofuran Complete
- FFKM, Fully fluorinated perfluoroelastomer PEEK, Polyetheretherketone Stainless steel
Specifications
Flow Restrictor | |
FFKM / PEEK / Stainless Steel | |
0.1 kg |